Learn More
MCR Safety Crews™ Tomahawk™ Safety Glasses
Catalog No. 189991907 Shop All MCR Safety Products
Click to view available options
Frame Color:
Black
Blue
Red, White, Blue
Lens Coating:
Scratch-resistant
Lens Tint:
Clear
Clear Mirror, Indoor/Outdoor
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy
The Tomahawk™ is an affordable alternative for providing comfort, protection and style. Features include a stylish wrap-around design and one-piece polycarbonate lens with integral sideshields.
  • High brow guard for extra protection and unimpeded view
  • Single lens design with integral side shields for greater orbital seal
  • Spatula temple with extendable adjustment
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Compliance
Spectacles meet ANSI Z87.1-1989 standards.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1-1989 safety standards
Lens Tint Clear
Frame Color Blue
Lens Material Polycarbonate
Frame Material Nylon
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.