Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety SensaTouch™ Latex Gloves

Catalog No. p-3659519 Shop All MCR Safety Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Natural rubber latex polymer gloves are comprised of biodegradable material that is inherently antimicrobial and antibacterial. Suitable for a variety of applications.

  • Disposable, Natural rubber latex
  • Reversible, rolled cuff
  • 11 mil thickness
  • Powder free, Textured grip
  • Double chlorinated
  • NFPA 1999 Certified
  • 12 in. extended length
  • Blue, Ambidextrous
  • Suitable for wide variety of applications including dentistry, laboratory technician, nursing home care, EMS, patient exams, first aid, painting and veterinarian

Specifications

Certifications/Compliance NFPA 1999, Medical Grade (Class 1 Medical Device)
Chemotherapeutic Rated No
Applications Healthcare, Laboratory, Industrial
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Laboratory/Cleanroom Use No
Latex Free No
Material Latex
Powdered Powder Free
Size Medium
Thickness (English) 11 mil
Thickness (Metric) 0.27 mm
FDA 510 (k) No
Model 5049-M
Series SensaTouch™
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.