Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety 5010 SensaTouch Vinyl Gloves

Catalog No. p-3659523
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Latex-free, powder free gloves suitable for a variety of applications including general medical, dental patient care, electronic assembly and food service.

  • Polyvinyl Chloride (PVC)
  • Powder free
  • Seamless, rolled cuffs
  • Ambidextrous
  • Medical Grade, Class 1 Medical Device
  • Complies with all relevant federal regulations for food contact

Specifications

Certifications/Compliance Medical Grade, Class 1 Medical Device, Complies with federal regulations for food contact
Chemotherapeutic Rated No
Applications Healthcare, Industrial
AQL 1.5
Color White
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 5 mil
Finger Thickness (Metric) 0.12 mm
Laboratory/Cleanroom Use No
Latex Free Yes
Material Vinyl
Powdered Powder Free
Size Small
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Model 5010S
Series SensaTouch™
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.