Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Eyeglass Cases

Catalog No. 19808736 Shop All MCR Safety Products
Click to view available options
Includes:
Belt Clip
Top flap, belt clip
This item is not returnable. View return policy
This item is not returnable. View return policy

Extends the life of protective eyewear

  • Belt clip
  • Vinyl material
  • Color: Black

Specifications

Description Eyeglass Case
Color Black
Product Type Eyewear Case
For Use With (Application) Extends the life of protective eyewear
Includes Belt Clip
Material Vinyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.