Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Cut Pro™ 9366 Aramid Fiber Gloves, PVC Dots
Shop All MCR Safety Products
Click to view available options
:
PVC dots on both sides
Description
- 7-gauge, Kevlar™ aramid fiber
- High comfort level
- Allows manual dexterity
- PVC dots on both sides
- Launderable
Specifications
Specifications
ANSI Abrasion Rating | ANSI 2 |
Coating Material | PVC Dots (2 Sides) |
Color | Yellow |
Cut Resistance Rating | ANSI A3 |
Material | Aramid Fiber, PVC |
Product Type | Cut-resistant Gloves |
Size | Large |
For Use With (Application) | air conditioning manufacturing, assembly line, cut protection, flame protection, food service, Glass handling, HVAC, Knife handling, light heat protection, warehousing |
Glove Cuff Style | Knit |
Hem Color | White |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction