Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety NXG™ Latex Coated Cotton/Polyester Work Gloves

Catalog No. 191203174B Shop All MCR Safety Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

A cotton/polyester seamless knit shell with latex-dipped palm and fingers for enhanced grip capabilities. Gloves are ideal for multi-industrial applications that require dexterity and comfort.

  • Seamless knit, 10-gauge cotton/polyester knit shell
  • Latex dip on the palm and fingertips for enhanced grip capabilities
  • Ideal for applications that require dexterity and comfort

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Gray
Cut Resistance Rating ANSI A2
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Polycotton
Product Type Seamless Knit Gloves
Size Medium
For Use With (Application) Automotive Assembly, Farming, Landscaping, Material Handling, and Carpentry
Glove Cuff Style Knit
Puncture Rating ASTM F2878 LEVEL 4
Series 9688
Yarn Gauge 10
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.