Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety White Nylon Inspection Gloves - Straight Thumb

Catalog No. p-3659624 Shop All MCR Safety Products
Click to view available options
Gender:
Men's
Women's
This item is not returnable. View return policy
This item is not returnable. View return policy

Smooth, thin and cool nylon inspection gloves are ideal for low lint applications. Wear alone or use as glove liners.

  • Breathable fabric helps keep hands cool
  • 100% Nylon for low lint applications
  • Comfortable and light weight
  • Straight thumb
  • Reversible

Specifications

Construction Jersey Knit
Description Low Lint
Color White
Hand Ambidextrous
Material Nylon
Product Type Inspection Gloves
Size Large
Style Reversible
Gender Men's
Glove Cuff Style Open
Packaging Type Polybag
Thumb Style Straight
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.