Learn More
MCR Safety Storm™Safety Glasses
Workers in the Storm™ can expect extreme comfort with this face-flattering protective eyewear from Crews™. Ratchet action temples combined with non- slip rubber head grips create a customized fit for virtually any head size.
$10.02 - $10.17
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
19-046-160
|
MCR Safety
ST1130 |
Green 3.0 |
Each for $10.17
Pack of 12 EA for $122.00
|
|
|||||
19-046-161
|
MCR Safety
ST1150 |
Green 5.0 |
Each for $10.02
Pack of 12 EA for $120.24
|
|
|||||
Description
Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection.- Available in a variety of frame colors and lens combinations
- Single wraparound dual focal point lens with integral side shields
- 5 position ratchet action bayonet temples
- Exclusive Duramass™ scratch-resistant coating
- Non-slip, soft temple material
Specifications
Safety Glasses | |
Black | |
Nylon |