Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCRS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597963
Description
MCRS1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus (PubMed:11948183). As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues (PubMed:20018852). Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity (PubMed:15044100). Binds to G-quadruplex structures in mRNA (PubMed:16571602). Binds to RNA homopolymer poly(G) and poly(U) (PubMed:16571602). [UniProt].
Specifications
MCRS1 | |
Polyclonal | |
Unconjugated | |
MCRS1 | |
58 kDa microspherule protein; C78274; cell cycle-regulated factor (78 kDa); cell cycle-regulated factor p78; ICP22BP; INO80 complex subunit J; INO80 complex subunit Q; INO80Q; Mcrs1; MCRS2; Microspherule protein 1; MSP58; nucleolar protein; P78 | |
Rabbit | |
Protein G | |
RUO | |
10445 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96EZ8 | |
MCRS1 | |
Recombinant Human Microspherule protein 1 protein (1-138AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction