Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCRS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119815 Supplier Invitrogen™ Supplier No. PA5119815
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: SH-SY5Y cell lysates, rat large intestine tissue, human liver tissue, human pancreas tissue, mouse testis tissue. Subcellular Location: Nucleus, Nucleolus, Cytoplasm.

Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus (PubMed:11948183). As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues (PubMed:20018852). Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity (PubMed:15044100). Binds to G-quadruplex structures in mRNA (PubMed:16571602). Binds to RNA homopolymer poly(G) and poly(U) (PubMed:16571602). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCRS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene MCRS1
Gene Accession No. Q96EZ8, Q99L90
Gene Alias 58 kDa microspherule protein; C78274; cell cycle-regulated factor (78 kDa); cell cycle-regulated factor p78; ICP22BP; INO80 complex subunit J; INO80 complex subunit Q; INO80Q; Mcrs1; MCRS2; Microspherule protein 1; MSP58; nucleolar protein; P78
Gene Symbols MCRS1
Host Species Rabbit
Immunogen Recombinant protein within human MCRS1 aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10445, 300222, 51812
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.