Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCRS1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MCRS1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MCRS1 Polyclonal specifically detects MCRS1 in Human, Mouse samples. It is validated for Western Blot.Specifications
MCRS1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Cell cycle-regulated factor p78, ICP22BP, INO80 complex subunit Q, INO80Q58 kDa microspherule protein, MCRS2INO80 complex subunit J, microspherule protein 1, MSP58cell cycle-regulated factor (78 kDa), P78 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 203-462 of human MCRS1 (NP_006328.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS with 50% glycerol, pH7.3. | |
10445 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title