Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572957
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572957 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572957 Supplier Invitrogen™ Supplier No. PA572957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCT1 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC16A1
Gene Accession No. P53985, P53986, P53987
Gene Alias AL022710; FLJ36745; HHF7; MCT; MCT 1; Mct1; MCT1D; MGC44475; monocarboxylate transporter; Monocarboxylate transporter 1; RATMCT1; RNMCT1; SLC16A1; SLC16A1 protein; solute carrier 16 (monocarboxylic acid transporter), member 1; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Symbols SLC16A1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Monocarboxylic acid transporter 1 protein (between residues 200-300).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20501, 25027, 6566
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.