Learn More
MCT1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304358AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.Specifications
MCT1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P53985 | |
SLC16A1 | |
Between 450 and 500 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
SLC16A1 | |
Multiple copies in a T-cell maligcies, Maligt T cell amplified sequence 1MCT1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6566 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.