Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCT1/SLC16A1 Antibody (882616), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8275H
Description
MCT1/SLC16A1 Monoclonal antibody specifically detects MCT1/SLC16A1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| MCT1/SLC16A1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6566 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 882616 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1) | |
| NS0 mouse myeloma cell line transfected with human MCT1/SLC16A1, Met1-Val500, Accession # P53985 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction