Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCT1/SLC16A1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159656AF405
Description
MCT1/SLC16A1 Polyclonal specifically detects MCT1/SLC16A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
MCT1/SLC16A1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated | |
FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1) | |
Rabbit | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
6566.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Alexa Fluor 405 | |
50 mM sodium borate with 0.05% sodium azide | |
SLC16A1 | |
A synthetic peptide made to an internal portion of the human Monocarboxylic acid transporter 1 protein (between residues 200-300) [UniProt P53985] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction