Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120845 Supplier Invitrogen™ Supplier No. PA5120845
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: DU145, HepG2, HL-60, 293T, rat brain. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: TIVECGPVLL GPPLAGKLVD LTGEYKYMYM SCGAIVVAAS VWLLIGNAIN YRLLAKERKE ENARQKTRES EPLSKSKHSE DVNVKVSNAQ SVTSERETNI.

Tissues with few or no mitochondria, such as erythrocytes and tumor cells, depend largely on glycolysis to generate ATP. The major end products of glycolysis, pyruvate and lactate, must be eliminated from these cells to enable continued glycolytic flux and prevent toxic effects. H+/monocarboxylate transporters mediate the transport of lactate and pyruvate. Human MCT2 has a high affinity for the transport of pyruvate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC16A7
Gene Accession No. O60669, Q63344
Gene Alias 4921534N07Rik; MCT 2; Mct2; monocarboxylate transporter; Monocarboxylate transporter 2; Slc16a7; solute carrier family 16 (monocarboxylate transporter), member 7; solute carrier family 16 (monocarboxylic acid transporters), member 7; solute carrier family 16 member 7; solute carrier family 16, member 7 (monocarboxylic acid transporter 2)
Gene Symbols SLC16A7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 379-478 of human SLC16A7 (NP_0012575511).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29735, 9194
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.