Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCT3/SLC16A8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159885 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159885 Supplier Novus Biologicals Supplier No. NBP159885
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCT3/SLC16A8 Polyclonal specifically detects MCT3/SLC16A8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MCT3/SLC16A8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O95907
Gene Alias MCT3MCT 3, monocarboxylate transporter 3, REMP, solute carrier 16 (monocarboxylic acid transporters), member 8, Solute carrier family 16 member 8, solute carrier family 16, member 8 (monocarboxylic acid transporter 3)
Gene Symbols SLC16A8
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC16A8(solute carrier family 16, member 8 (monocarboxylic acid transporter 3)) The peptide sequence was selected from the middle region of SLC16A8. Peptide sequence RAFAVYAVTKFLMALGLFVPAILLVNYAKDAGVPDTDAAFLLSIVGFVDI The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 55 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23539
Test Specificity Expected identity based on immunogen sequence: Chicken: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.