Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCT8/SLC16A2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232109AF594
Description
MCT8/SLC16A2 Polyclonal antibody specifically detects MCT8/SLC16A2 in Human, Mouse samples. It is validated for Western BlotSpecifications
MCT8/SLC16A2 | |
Polyclonal | |
Western Blot | |
DXS128, DXS128E, MCT 7, MCT 8, solute carrier family 16, member 2 (monocarboxylic acid transporter 8), XPCT | |
The immunogen this antibody was made to, maps to a region between residue 250 to 300 of human Monocarboxylate Transporter 8 using the numbering given in entry NP_006508.2 (GeneID 6567). | |
0.1 mL | |
Cell Biology, Endocrinology, Neuroscience, Signal Transduction | |
6567 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction