Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCTP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569310 Supplier Invitrogen™ Supplier No. PA569310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Yeast: 83%; Zebrafish: 100%.

MCTP1 belongs to the MCTP family.It contains 3 C2 domains. MCTP1 is a multi-pass membrane protein. It binds calcium via the C2 domains in absence of phospholipids. The function of MCTP1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCTP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MCTP1
Gene Accession No. E9PV86
Gene Alias 2810465F10Rik; LOW QUALITY PROTEIN: multiple C2 and transmembrane domain-containing protein 1; MCTP1; Mctp1-ps1; multiple C2 and transmembrane domain containing 1; multiple C2 and transmembrane domain-containing protein 1; multiple C2 and transmembrane domain-containing protein 1 pseudogene; multiple C2 and transmembrane domain-containing protein 1-like; Multiple C2 and transmembrane domain-containing protein 1-like protein; multiple C2 domains, transmembrane 1; multiple C2 domains, transmembrane 1, pseudogene 1; multiple C2-domains with two transmembrane regions 1; RGD1305199
Gene Symbols MCTP1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse MCTP1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78771
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.