Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCTP1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA553893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553893 Supplier Thermo Scientific Supplier No. PA553893

Rabbit Polyclonal Antibody

Immunogen sequence: VFDYDFGLQD DFMGSAFLDL TQLELNRPTD VTLTLKDPHY PDHDLGIILL SVILTPKEGE SRDVTMLMRK SWKRSSKFQT QSLRLSDLHR KSHLWRGIVS ITLIEGRDLK AMDSNGLSDP YVKFRLGH Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

MCTP1 belongs to the MCTP family.It contains 3 C2 domains. MCTP1 is a multi-pass membrane protein. It binds calcium via the C2 domains in absence of phospholipids. The function of MCTP1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCTP1
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCTP1
Gene Accession No. Q6DN14
Gene Alias 2810465F10Rik; LOW QUALITY PROTEIN: multiple C2 and transmembrane domain-containing protein 1; MCTP1; Mctp1-ps1; multiple C2 and transmembrane domain containing 1; multiple C2 and transmembrane domain-containing protein 1; multiple C2 and transmembrane domain-containing protein 1 pseudogene; multiple C2 and transmembrane domain-containing protein 1-like; Multiple C2 and transmembrane domain-containing protein 1-like protein; multiple C2 domains, transmembrane 1; multiple C2 domains, transmembrane 1, pseudogene 1; multiple C2-domains with two transmembrane regions 1; RGD1305199
Gene Symbols MCTP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCTP1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79772
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.