Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCTP2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298643PCP
Description
MCTP2 Polyclonal antibody specifically detects MCTP2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
MCTP2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
FLJ11175, FLJ33303, MGC111402, multiple C2 and transmembrane domain-containing protein 2, multiple C2 domains, transmembrane 2, multiple C2-domains with two transmembrane regions 2 1 | |
Produced in rabbits immunized with E. coli-derived Human MCTP2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55784 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction