Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCTP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567109 Supplier Invitrogen™ Supplier No. PA567109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHRWSNRKRLS ASKSSLIRNL RLSESLKKNQ LWNGIISITL LEGKNVSGGS MTEMFVQLKL GDQRYKSKTL CKSANPQW Highest antigen sequence identity to the following orthologs - mouse 96%, rat 94%.

Might play a role in the development of cardiac outflow tract. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCTP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCTP2
Gene Accession No. Q6DN12
Gene Alias Gm489; MCTP2; multiple C2 and transmembrane domain containing 2; multiple C2 and transmembrane domain-containing protein 2; multiple C2 domains, transmembrane 2; multiple C2-domains with two transmembrane regions 2 1; RGD1562967
Gene Symbols MCTP2
Host Species Rabbit
Immunogen Recombinant Human MCTP2. Recombinant protein control fragment (Product #RP-105506).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55784
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.