Learn More
MCTP2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305695AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function of this protein remains unknown.Specifications
MCTP2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6DN12 | |
MCTP2 | |
Between 100 and 150 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
MCTP2 | |
multiple C2 and transmembrane domain-containing protein 2, multiple C2-domains with two transmembrane regions 2 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55784 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.