Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCTS1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen MCTS1
Concentration 1 mg/mL
Dilution Western Blot 1:500
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018175
SDP
View Documents
Novus Biologicals
NB500128SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB500128
SDP
View Documents
Novus Biologicals
NB500128
0.1 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

MCTS1 Polyclonal specifically detects MCTS1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MCTS1
Western Blot 1:500
Polyclonal
Rabbit
Cardiovascular Biology
PBS with 0.02% Sodium Azide
28985
Partial residue sequence of the 20 kDa MCT1 protein
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Mouse, Human
malignant T cell amplified sequence 1, Malignant T-Cell-Amplified Sequence 1, MCT-1, Multiple Copies T-Cell Malignancies
MCTS1
IgG
Affinity Purified
This is specific for MCT1.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.