Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCTS1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299158MFV500
Description
MCTS1 Polyclonal antibody specifically detects MCTS1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MCTS1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
malignant T cell amplified sequence 1, Malignant T-Cell-Amplified Sequence 1, MCT-1, Multiple Copies T-Cell Malignancies | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MCTS1 | |
0.1 mL | |
Cardiovascular Biology | |
28985 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction