Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCTS1 Antibody (OTI 2G2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272602AF750
Description
MCTS1 Monoclonal specifically detects MCTS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MCTS1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
MCTS1 | |
Recombinant protein fragment corresponding to amino acids 1-181 of human MCTS1 (NP_054779) produced in E.coli. | |
Cardiovascular Biology | |
28985 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI 2G2 | |
Western Blot, Immunohistochemistry | |
malignant T cell amplified sequence 1, Malignant T-Cell-Amplified Sequence 1, MCT-1, Multiple Copies T-Cell Malignancies | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction