Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCTS1 Monoclonal Antibody (OTI 5B5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502506S


Description
MCTS1 Monoclonal antibody specifically detects MCTS1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
MCTS1 | |
Monoclonal | |
0.77 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N9V6 | |
Mouse | |
Affinity chromatography | |
RUO | |
79998 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI 5B5 | |
Unconjugated | |
ANKRD53 | |
1500019M23Rik; malignant T cell amplified sequence 1; malignant T cell-amplified sequence 1; malignant T-cell amplified sequence 1; malignant T-cell-amplified sequence 1; MCT1; MCT-1; MCTS1; MCTS1, re-initiation and release factor; multiple copies in T-cell lymphoma-1; multiple copies T-cell malignancies; Multiple copies T-cell malignancies 1 | |
Human recombinant protein fragment corresponding to amino acids 1-300 of human ANKRD53 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction