Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCTS1 Monoclonal Antibody (OTI2F9)
GREENER_CHOICE

Catalog No. PIMA525417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525417 Supplier Invitrogen™ Supplier No. MA525417
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCTS1 Monoclonal Antibody for Western Blot, IHC (P)

MCTS1 play a role in cell cycle regulation; decreases cell doubling time and anchorage-dependent growth; shortens the duration of G1 transit time and G1/S transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCTS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F9
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MCTS1
Gene Accession No. Q9ULC4
Gene Alias 1500019M23Rik; malignant T cell amplified sequence 1; malignant T cell-amplified sequence 1; malignant T-cell amplified sequence 1; malignant T-cell-amplified sequence 1; MCT1; MCT-1; MCTS1; MCTS1, re-initiation and release factor; multiple copies in T-cell lymphoma-1; multiple copies T-cell malignancies; Multiple copies T-cell malignancies 1
Gene Symbols MCTS1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-181 of MCTS1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28985
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.