Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCTS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581293 Supplier Invitrogen™ Supplier No. PA581293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MCT1/MCTS1.

MCTS1 play a role in cell cycle regulation; decreases cell doubling time and anchorage-dependent growth; shortens the duration of G1 transit time and G1/S transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCTS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MCTS1
Gene Accession No. Q9ULC4
Gene Alias 1500019M23Rik; malignant T cell amplified sequence 1; malignant T cell-amplified sequence 1; malignant T-cell amplified sequence 1; malignant T-cell-amplified sequence 1; MCT1; MCT-1; MCTS1; MCTS1, re-initiation and release factor; multiple copies in T-cell lymphoma-1; multiple copies T-cell malignancies; Multiple copies T-cell malignancies 1
Gene Symbols MCTS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human MCTS1/Malignant T cell amplified sequence 1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.