Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCTS1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142973
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isosoforms (NP_054779.1; NP_001131026.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
MCTS1 play a role in cell cycle regulation; decreases cell doubling time and anchorage-dependent growth; shortens the duration of G1 transit time and G1/S transition.
Specifications
MCTS1 | |
Polyclonal | |
Unconjugated | |
MCTS1 | |
1500019M23Rik; malignant T cell amplified sequence 1; malignant T cell-amplified sequence 1; malignant T-cell amplified sequence 1; malignant T-cell-amplified sequence 1; MCT1; MCT-1; MCTS1; MCTS1, re-initiation and release factor; multiple copies in T-cell lymphoma-1; multiple copies T-cell malignancies; Multiple copies T-cell malignancies 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
28985 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9ULC4 | |
MCTS1 | |
Peptide with sequence DEKENVSNCIQLKTS (aa 6-20). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction