Learn More
Cytiva Whatman™ Replacement Teflon Support Screen for MD050 Series Pressure Filtration Manifolds
Shop All Cytiva ProductsDescription
- Product Type: Support Screen
- Model: MD050/0/03
- Material: Teflon
- For Use With (Application): Pressure Filtration
- For Use With (Equipment): MD050 Series Pressure Filtration Manifolds
Specifications
Specifications
| Disposable | Reusable |
| For Use With (Equipment) | MD050 Series Pressure Filtration Manifolds |
| Material | Teflon |
| Sterility | Non-sterile |
| For Use With (Application) | Pressure Filtration |
| Model | MD050/0/03 |
| Quantity | 1 Ea. |
| Series | MD 050 |
| Type | Support Screen |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.