Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MD-1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Specifications
Antigen | MD-1/LY86 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Polyclonal |
Description
MD-1/LY86 Polyclonal specifically detects MD-1/LY86 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
MD-1/LY86 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
Ly-86, LY86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
LY86 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
RUO | |
O95711 | |
9450 | |
Mouse myeloma cell line NS0-derived recombinant human MD-1 Gly21-Ser162 Accession # O95711 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human MD-1 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse MD-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title