Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MD-1/LY86 Antibody (416), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290387MFV500
Description
MD-1/LY86 Monoclonal antibody specifically detects MD-1/LY86 in Mouse samples. It is validated for ELISASpecifications
MD-1/LY86 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
416 | |
ELISA | |
Ly-86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
Rabbit immunized with purified, recombinant Mouse MD-1/LY86 (Met 1-Ser 162, Accession#: NP_034875.1) | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Immunology | |
9450 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction