Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MD-1 Monoclonal Antibody (MD113), Biotin

Catalog No. 01676497
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-497 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-497 Supplier Invitrogen™ Supplier No. MA140041

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

MA1-40041 detects MD-1 from mouse samples. MA1-40041 has been successfully used in flow cytometry and functional assays applications. The MA1-40041 immunogen is mouse MD-1 extracellular protein.

MD-1 is an extracellular protein associated with the extracellular domain of RP105.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MD-1
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone MD113
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene LY86
Gene Accession No. O88188
Gene Alias dJ80N2.1; Ly86; Ly-86; Lymphocyte antigen 86; MD1; MD-1; MD-1, RP105-associated; MMD-1; Protein MD-1; RP1-80N2.1
Gene Symbols LY86
Host Species Rat
Immunogen Murine MD-1 extracellular protein.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17084
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.