Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MD-1 Rat anti-Mouse, PE, Clone: MD113 (also known as MD-113), BD Phrmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 569973
Description
MD-1 Monoclonal antibody specifically detects MD-1 in Mouse samples. It is validated for Flow cytometry
Specifications
MD-1 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
17084 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2b κ |
Flow Cytometry | |
MD113 (also known as MD-113) | |
PE | |
Ly86; MD-1; MD1; ly-86; lymphocyte antigen 86 | |
Mouse RP105/MD-1 Transfected Cells | |
25 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction