Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MD-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177201
Description
MD-2 Polyclonal specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
MD-2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 0.5-1 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 2-2.5 ug/mL, Immunocytochemistry/Immunofluorescence 10-20 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml | |
NP_056179 | |
LY96 | |
Antibody was raised against a peptide corresponding to 13 amino acids near the center of human MD-2. The immunogen is located within the last 50 amino acids of MD-2. Amino Acid Squence: CSFKGIKFSKGKYK | |
0.1 mg | |
Signal Transduction | |
23643 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction