Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MD-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$427.50
Specifications
Antigen | MD-2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5-10 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056655
![]() |
Novus Biologicals
NB10056655 |
0.1 mg |
Each of 1 for $427.50
|
|
|||||
Description
MD-2 Polyclonal specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MD-2 | |
Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5-10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
NP_056179 | |
23643 | |
A full length recombinant protein was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS with 0.05% Sodium Azide | |
ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
LY96 | |
IgG | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title