Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MD-2 Monoclonal Antibody (18H10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA533351
Description
Product is supplied at 1 mL (100 μL/mL). Application Notes: This antibody can be used to inhibit LPS binding to MD-2 in vitro. For functional assays, dilutions have to be made according to the amounts of MD-2 to be inactivated.
MD-2 is a gene that encodes a protein involved in the recognition of bacterial lipopolysaccharides (LPS) by the immune system. It is located on chromosome 8 and is part of a larger family of genes that encode for similar proteins. The MD-2 protein forms a complex with Toll-like receptor 4 (TLR4) to recognize and respond to LPS, triggering an immune response. Mutations or variations in the MD-2 gene have been associated with various diseases and conditions, including sepsis and inflammatory disorders.
Specifications
MD-2 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and no preservative | |
Q9Y6Y9 | |
LY96 | |
TLR4/MD-2 expressing CHO cells/ chimeric TLR4/MD-2 fusion protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Functional Assay | |
18H10 | |
Unconjugated | |
LY96 | |
Esop1; ESOP-1; LPS co-receptor MD-2; LY96; ly-96; Lymphocyte antigen 96; MD2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2 | |
Mouse | |
Protein G | |
RUO | |
23643 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction