Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MDA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102605
Description
Antibody detects endogenous levels of total IFIH1.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein. Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.
Specifications
MDA5 | |
Polyclonal | |
Unconjugated | |
IFIH1 | |
9130009C22Rik; AGS7; CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; helicard; Helicase with 2 CARD domains; Hlcd; IDDM19; IFIH1; interferon induced with helicase C domain 1; interferon induced with helicase C domain protein 1; interferon induced, with helicase C domain 1; interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA5; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RGD1560496; RH116; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116; SGMRT1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
499801, 64135, 71586 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8R5F7, Q9BYX4 | |
IFIH1 | |
A synthesized peptide derived from human IFIH1(Accession Q9BYX4), corresponding to amino acid residues M971-F1021. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction