Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MDC Monoclonal Antibody (57226)
Mouse Monoclonal Antibody
$643.00 - $976.50
Specifications
Antigen | MDC |
---|---|
Clone | 57226 |
Concentration | 0.5 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Neutralization, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA523754
|
Invitrogen™
MA523754 |
500 μg |
Each for $976.50
|
|
|||||
PIMA523835
|
Invitrogen™
MA523835 |
100 μg |
Each for $643.00
|
|
|||||
Description
In ELISAs, this antibody does notcross-react with recombinant mouse (rm) CCL17, rhCCL17, or rhCCL21. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.Specifications
MDC | |
0.5 mg/mL | |
ELISA, Neutralization, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 5% trehalose and No Preservative | |
O00626 | |
6367 | |
E. coli-derived recombinant human CCL22/MDC Gly25-Gln93. | |
Antibody |
57226 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Lyophilized | |
IgG2b | |
Human | |
CCL22 | |
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
CCL22 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title