Learn More
MDC Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$2175.00
Specifications
Antigen | MDC |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-271-8710
![]() |
Invitrogen™
500P107BT500UG |
500 μg |
Each of 1 for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: GPYGANMEDS VCCRDYVRYR LPLRVVKHFY WTSDSCPRPG VVLLTFRDKE ICADPRVPWV KMILNKLSQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MDC (CCL22). Anti-Human MDC (CCL22)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human MDC (CCL22) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MDC (CCL22) (500-P107) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MDC (CCL22). Western Blot: To detect hMDC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMDC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P107BT-1MG will be provided as 2 x 500 μg
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.Specifications
MDC | |
ELISA, Western Blot | |
Biotin | |
Rabbit | |
Human | |
O00626 | |
6367 | |
E.coli-derived Recombinant Human MDC (CCL22) (69 a.a.). | |
Antigen affinity chromatography | |
CCL22 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
CCL22 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.