Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDC1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB275102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB275102 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB275102 Supplier Novus Biologicals Supplier No. NB100397AF700
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDC1 Polyclonal antibody specifically detects MDC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MDC1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp781A0122, Em:AB023051.5, KIAA0170MGC166888, mediator of DNA damage checkpoint 1, mediator of DNA damage checkpoint protein 1, mediator of DNA-damage checkpoint 1, NFBD1homologue to Drosophila photoreceptor protein calphotin, Nuclear factor with BRCT domains 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residues 1375 and 1425 of human Mediator of DNA Damage Checkpoint 1 using the numbering given in entry NP_055456.1 (GeneID 9656).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9656
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.