Learn More
Invitrogen™ MDC1 Polyclonal Antibody

Description
Suggested positive control: Jurkat whole cell lysate. Immunogen sequence: RQESDKNGES ASPERDRE.
Specifications
Specifications
| Antigen | MDC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | MDC1 |
| Gene Accession No. | Q14676, Q5PSV9, Q5U2M8 |
| Gene Alias | 6820401C03; AA413496; DAAP-285E11.7; DKFZp781A0122; homologue to Drosophila photoreceptor protein calphotin; KIAA0170; MDC1; mediation of DNA damage checkpoint 1; mediator of DNA damage checkpoint 1; mediator of DNA damage checkpoint protein 1; mediator of DNA-damage checkpoint 1; MGC166888; mKIAA0170; NFBD1; nuclear factor with BRCT domains 1 |
| Gene Symbols | MDC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.