Learn More
3B Scientific™ MDF™ Calibra™ Pro Aneroid Sphygmomanometer
SDPSCI_ED
Catalog No. S00213
Click to view available options
Color:
Navy Blue
This item is not returnable. View return policy
This item is intended for educational purposes only.
This item is not returnable. View return policy
This item is intended for educational purposes only.
The MDF™ Calibra™ Pro Aneroid Sphygmomanometer is ideal for medical professionals in all settings. Stress-tested over 3,000 times and calibrated 5 times, it is one of the most durable, accurate, and dependable blood pressure measurement devices on the market.
  • Gauge:
    • A precise, certified 300 mmHg manometer attains the accuracy of ±3 mmHg without pin stop, and features an easily identifiable high-contrast dial.
    • The die-cast zinc manometer housing is finished in textured baked enamel and outfitted with a heavy-duty clip so it can be easily attached to the gauge holder on the cuff.
  • Cuff & Inflation Bag:
    • Resistant to abrasion, chemicals (chlorine and peroxide), and moisture, the adult Velcro™ cuff is constructed of high-molecular polymer nylon.
    • Sized to meet American Heart Association recommendations, the artery indicator label and index range further ensure proper cuffing for correct arterial compression.
    • The double tube seamless inflation bag is constructed of crack-resistant, non-sticking, high-density hypoallergenic latex-free PVC.
    • The universal bladder tube enables compatible cuff exchange with all MDF™ and other major brands of blood pressure systems.
  • Inflation Bulb & Valve:
    • Chrome-plated brass screw-type valve delivers precise deflation control rate. The inflation bulb is constructed of crack-resistant, non-sticking, high-density hypoallergenic latex-free PVC.
  • Included Accessories: Nylon zippered carrying case with Velcro™ side pocket, ID tag.
Specifications
Product Type Aneroid Sphygmomanometer
Color Navy Blue
Includes Nylon Zippered Carrying Case With Velcro™ Side Pocket, ID Tag
Weight (English) 0.96 lb.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.