Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDFI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562108 Supplier Invitrogen™ Supplier No. PA562108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPNDSGHPSE LGGTRRAGNG ALGGPKAHRK LQTHPSLASQ GSKKSKSSSK STTSQIPLQA QEDCC Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 72%.

MDFI is a transcription factor that negatively regulates other myogenic family proteins. Studies of the mouse homolog, I-mf, show that it interferes with myogenic factor function by masking nuclear localization signals and preventing DNA binding. Knockout mouse studies show defects in the formation of vertebrae and ribs that also involve cartilage formation in these structures. MDFI an also interfere with the DNA-binding activity of Myod family members and plays an important role in trophoblast and chondrogenic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDFI
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mdfi
Gene Accession No. Q99750
Gene Alias I-mf; I-mfa; inhibitor of MyoD family a; MDFI; MyoD family inhibitor; myogenic repressor I-mf; RGD1560271
Gene Symbols Mdfi
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MDFI. Recombinant protein control fragment (Product #RP-101208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4188
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.