Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDFIC Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550706

Catalog No. PA550706


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total MDFIC protein.

MDFIC is a member of a family of proteins characterized by a specific cysteine-rich C-terminal domain, which is involved in transcriptional regulation of viral genome expression. Alternative translation initiation from an upstream non-AUG (GUG), and an in-frame, downstream AUG codon, results in the production of two isoforms, p40 and p32, respectively, which have different subcellular localization; p32 is mainly found in the cytoplasm, whereas p40 is targeted to the nucleolus. Both isoforms have transcriptional regulatory activity that is attributable to the cysteine-rich C-terminal domain.This gene product is a member of a family of proteins characterized by a specific cysteine-rich C-terminal domain, which is involved in transcriptional regulation of viral genome expression. Alternative translation initiation from an upstream non-AUG (GUG), and an in-frame, downstream AUG codon, results in the production of two isoforms, p40 and p32, respectively, which have different subcellular localization; p32 is mainly found in the cytoplasm, whereas p40 is targeted to the nucleolus. Both isoforms have transcriptional regulatory activity that is attributable to the cysteine-rich C-terminal domain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MDFIC
Polyclonal
Unconjugated
MDFIC
HIC; I-mfa domain-containing protein; MDFIC; MDFIC (HGNC:28870); MyoD family inhibitor domain containing; MyoD family inhibitor domain containing protein; myoD family inhibitor domain-containing protein
Rabbit
Antigen affinity chromatography
RUO
29969
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9P1T7
MDFIC
Synthetic peptide corresponding to a region derived from internal residues of human MyoD family inhibitor domain containing.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.