Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217246
Description
MDG1 Polyclonal specifically detects MDG1 in Human, Mouse samples. It is validated for Western Blot.Specifications
MDG1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
DKFZp564F1862, DnaJ (Hsp40) homolog, subfamily B, member 9, dnaJ homolog subfamily B member 9, endoplasmic reticulum DnaJ homolog 4, ERdj4, Mdg-1, MDG1MSTP049, Microvascular endothelial differentiation gene 1 protein, MST049 | |
Rabbit | |
26 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (100%), Rat (100%), Xenopus laevis (100%), Bovine (100%), Xenopus tropicalis (100%), Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.32mg/mL | |
Western Blot 1:500-1:3000 | |
Q9UBS3 | |
DNAJB9 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MDG1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
4189 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction