Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDGA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547691
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547691 Supplier Invitrogen™ Supplier No. PA547691
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human MDGA2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MDGA1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene MDGA1
Gene Accession No. Q8NFP4
Gene Alias FLJ45018; glycosyl-phosphatidyl-inositol-MAM; glycosylphosphatidylinositol-MAM; GPI and MAM protein; GPIM; MAM domain containing glycosylphosphatidylinositol anchor 1; MAM domain-containing glycosylphosphatidylinositol anchor protein 1; MAM domain-containing protein 3; MAMDC3; MDGA1
Gene Symbols MDGA1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human MDGA1 Gln19-Gly931.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266727
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.