Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDGA2 Antibody (803732), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5184PEATT594
Description
MDGA2 Monoclonal antibody specifically detects MDGA2 in Human, Rat samples. It is validated for Flow CytometrySpecifications
MDGA2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
161357 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
803732 | |
Flow Cytometry | |
MAM domain containing glycosylphosphatidylinositol anchor 2 | |
Mouse myeloma cell line NS0-derived recombinant human MDGA2, Gln21-Asp934, Accession # Q7Z553 | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction