Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDGA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545835 Supplier Invitrogen™ Supplier No. PA545835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TATRNTKYTP NTGPNADRSG SKEGFYMYIE TSRPRLEGEK ARLLSPVFSI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

MDGA2 may involved in cell-cell interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDGA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MDGA2
Gene Accession No. Q7Z553
Gene Alias 6720489L24Rik; 9330209L04Rik; acrodysplasia; Adp; c14_5286; MAM domain containing 1; MAM domain containing glycosylphosphatidylinositol anchor 2; MAM domain-containing glycosylphosphatidylinositol anchor protein 2; MAM domain-containing protein 1; MAMDC1; Mdga2; UNQ8188/PRO23197
Gene Symbols MDGA2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MDGA2 (aa 773-822).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161357
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.